missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FIP1L1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541914
This item is not returnable.
View return policy
Description
Peptide sequence: EYAERGYERH RASREKEERH RERRHREKEE TRHKSSRSNS RRRHESEEGD Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 86%.
FIP1L1 is a component of the cleavage and polyadenylation specificity factor (CPSF) complex that plays a key role in pre-mRNA 3'-end formation, recognizing the AAUAAA signal sequence and interacting with poly(A) polymerase and other factors to bring about cleavage and poly(A) addition. FIP1L1 contributes to poly(A) site recognition and stimulates poly(A) addition. It binds to U-rich RNA sequence elements surrounding the poly(A) site. FIP1L1 may act to tether poly(A) polymerase to the CPSF complex.
Specifications
| FIP1L1 | |
| Polyclonal | |
| Unconjugated | |
| FIP1L1 | |
| 1300019H17Rik; DKFZp586K0717; Factor interacting with PAP; factor interacting with PAPOLA and CPSF1; FIP1; FIP1 like 1; FIP1 like 1 (S. cerevisiae); FIP1 like 1b (S. cerevisiae); FIP1L1; FIP1L1 cleavage and polyadenylation specific factor subunit; fip1l1b; FIP1-like 1 protein; FLJ33619; hFip1; Pre-mRNA 3'-end-processing factor FIP1; pre-mRNA 3'-end-processing factor FIP1; LOW QUALITY PROTEIN: pre-mRNA 3'-end-processing factor FIP1; rearranged in hypereosinophilia; RHE; Rje; wu:fd36f11; zgc:103421 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 81608 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6UN15 | |
| FIP1L1 | |
| Synthetic peptide directed towards the C-terminal of human FIP1L1 (aa 509-558). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction