Learn More
Thermo Scientific™ Finnpipette Stepper Pipette
Product Code. 11863011
Click to view available options
Volume (Metric):
10 to 5000 μL
Unit Size:
Each
Product Code. 11863011
Brand: Thermo Scientific™ 4540000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Rapidly dispense up to 45 times in succession without refilling. This lightweight, easy to use repeater pipetter is designed for one-handed dispensing.
Rapidly dispense up to 45 times in succession without refilling. The Thermo Scientific™ Finnpipette™ Stepper Pipette is a lightweight, easy to use repeater pipette ergonomically designed for one-handed dispensing. Operating on the positive displacement principle, the Finnpipette Stepper is ideally suited for work with aggressive and viscous liquids.
Features:
  • Positive displacement design ensures accurate results, even for viscous, volatile, or other problematic liquids
  • Lightweight, ergonomic handle and wide finger rest minimize operator hand strain
  • Operation is simple—just attach the appropriate tip size, fill and prime the tip, and dial the selected volume
  • Volume is easy to set, with the aid of the convenient volume-setting chart located on the handle
Ordering Information:
Universal Stepper handle is also available with an eight-channel module. For convenient storage, pipetter stand is available (Cat. No. 9420290).
Includes:
Unit comes with one adapter (nonsterile) necessary for use with 25 or 50mL tips.
Requires:
Uses the Thermo Scientific Finntip™ Stepper tip, a plunger-style tip available in nonsterile or sterile (individually wrapped) form. This tip comes in seven sizes from 0.5 to 50mL, each with five different delivery volumes, for an overall dispensing range of 10 to 5000μL. Only the disposable tip comes in contact with the sample, eliminating the risk of contaminating the pipetter body.
TRUSTED_SUSTAINABILITY
Specifications
Description Finnpipette Stepper Pipetter
For Use With FinnTip
Model Stepper
Number of Channels 1
Type Stepper Pipette
Volume (Metric) 10 to 5000 μL