missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Finnpipette™ Stepper Pipette Adapters for 25 and 50mL tips

Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 10
Pack of 5
Description
Combine these Thermo Scientific™ Finnpipette™ Stepper Pipette Adapters for 25 and 50mL tips with your pipette. Choose sterile or nonsterile.
Specifications
Specifications
| Description | For 25 or 50mL Finnitip Stepper tips |
| For Use With | FinnTip Stepper Tip (25, 50 mL) |
| Sterility | Non-sterile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction