Learn More
Thermo Scientific™ Finnpipette™ Multistepper Pipette
Description
Reduce the risk of error and increase productivity in immunoassay applications with the Thermo Scientific™ Finnpipette™ Multistepper Pipette. This lightweight, easy-to-use repeater pipette provides simultaneous dispensing in eight channels. Designed especially for work with microplates, the Finnpipette Multistepper helps speed up routine laboratory procedures and is easy to use — simply attach, fill, and prime the module, then dial the selected volume.
Design
- Universal handle also fits the seven single-channel Thermo Scientific™ Finntip™ Steppers
- Lightweight construction minimizes hand fatigue during repetitive dispensing
- Separate tip ejector for safe and effortless tip disposal
- Wide finger rest minimizes operator hand strain
- Module rotates a full 360°, optimizing efficiency for both right or left-handed users
Compatible with:
Accepts Finntip Multistepper 1500μL tips. They are extra long, with more air space between the liquid and tip cone to prevent contamination.
Specifications
Specifications
| Description | Handle and module |
| For Use With | FinnTip™ Multistepper 1500 μL Tip |
| Number of Channels | 8 |
| Type | Multistepper Pipette |
| Volume (Metric) | 50 to 250 μL |