Learn More
Thermo Scientific™ Finnpipette™ F1 GLP Kits
Description
Access everything needed for pipetting in these all-inclusive kits. Thermo Scientific™ Finnpipette™ F1 GLP (Good Laboratory Pipetting) Kit includes state-of-the-art Finnpipette F1 pipetters which offer superior performance and unique features such as adjustable finger rest and antimicrobial surface for user comfort.
Features:
- Adjustable finger rest for increased ergonomicss and comfortability
- Set and forget pipetting button for easy and secured volume adjustment
- Antimicrobial surface for better protection
- Large easy to read display for better vision ergonomics
- Soft-touch tip ejection for light end effortless tip ejection
Kit 1 includes:
- Three Thermo Scientific Finnpipette Pipetters from 1 to 1000μL:
- Finnpipette F1, 1-10μL
- Finnpipette F1, 10-100μL
- Finnpipette F1, 100-1000μL
- Finntip Flex tips: 10, 1x96; 200, 1x96; 1000, 1x96
- F-stand; F1 pen; F1 brochure
- Good Laboratory Pipetting Guide
- Reagent reservoir demo pack
Kit 2 includes:
- Four Thermo Scientific Finnpipette Pipetters from 0.2 to 1000μL:
- Finnpipette F1, 0.2-2μL
- Finnpipette F1, 2-20μL univ
- Finnpipette F1, 20-200μL
- Finnpipette F1, 100-1000μL
- Finntip Flex tips: 10, 1x96; 200, 2x96; 1000, 1x96
- F-stand; F1 pen; F1 brochure
- Good Laboratory Pipetting Guide
- Reagent reservoir demo pack
Kit 3 includes:
- Three Thermo Scientific Finnpipette Pipetters from 10 to 10000μL:
- Finnpipette F1, 10-100μL
- Finnpipette F1, 100-1000μL
- Finnpipette F1, 1000-10000μL
- Finntip Flex tips: 200, 1x96; 1000, 1x96; FT 10mL, 1x24 F-stand; F1 pen; F1 brochure
- Good Laboratory Pipetting Guide
- Reagent reservoir demo pack
Kit 4 includes:
- Three Thermo Scientific Finnpipette Pipetters from 10 to 1000μL:
- Finnpipette F1, 2-20μL
- Finnpipette F1, 20-200μL
- Finnpipette F1, 100-1000μL
- Flex tips: (2) 200μL, 10x96; 100-1000μL, 10x96
- F-stand; F1 pen; F1 brochure
- Good Laboratory Pipetting Guide
- Reagent reservoir demo pack
Ordering Information:
Good Laboratory Pipetting kit also includes samples of Finntip Flex for maximum performance and best ergonomics. The kit contains a Good Laboratory Pipetting guide offering a extensive information of pipetting and factors influencing to the pipetting results.
Compatible with:
Finntip, ART tip, HLT tip
Specifications
Specifications
| Compatible Tips | Finntip Flex tips: 200; 1000; FT 10mL |
| Description | Kit 3: Finnpipette F1 10-100μL, 100-1000μL and 1-10mL |
| Color | Assorted |
| For Use With | Finntip: Flex Tips: 200, 1000 |
| Model | F1 |
| Number of Channels | 1 |
| Product Line | Finnpipette™ F1 |
| Type | GLP Kit |
| Volume (Metric) | 10 to 10,000 μL |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.