missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Finnpipette™ F1 GLP Kits

Product Code. 11805850
Click to view available options
Volume (Metric):
0.2 to 1000 μL
1 to 1000 μL
10 to 10,000 μL
2 to 1000 μL
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 11805850

Brand: Thermo Scientific™ 4700850

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Access everything needed for pipetting in these all-inclusive kits featuring state-of-the-art pipetters that maximize reliability and comfort.

Access everything needed for pipetting in these all-inclusive kits. Thermo Scientific™ Finnpipette™ F1 GLP (Good Laboratory Pipetting) Kit includes state-of-the-art Finnpipette F1 pipetters which offer superior performance and unique features such as adjustable finger rest and antimicrobial surface for user comfort.

Features:

  • Adjustable finger rest for increased ergonomicss and comfortability
  • Set and forget pipetting button for easy and secured volume adjustment
  • Antimicrobial surface for better protection
  • Large easy to read display for better vision ergonomics
  • Soft-touch tip ejection for light end effortless tip ejection

Kit 1 includes:

  • Three Thermo Scientific Finnpipette Pipetters from 1 to 1000μL:
  • Finnpipette F1, 1-10μL
  • Finnpipette F1, 10-100μL
  • Finnpipette F1, 100-1000μL
  • Finntip Flex tips: 10, 1x96; 200, 1x96; 1000, 1x96
  • F-stand; F1 pen; F1 brochure
  • Good Laboratory Pipetting Guide
  • Reagent reservoir demo pack

Kit 2 includes:

  • Four Thermo Scientific Finnpipette Pipetters from 0.2 to 1000μL:
  • Finnpipette F1, 0.2-2μL
  • Finnpipette F1, 2-20μL univ
  • Finnpipette F1, 20-200μL
  • Finnpipette F1, 100-1000μL
  • Finntip Flex tips: 10, 1x96; 200, 2x96; 1000, 1x96
  • F-stand; F1 pen; F1 brochure
  • Good Laboratory Pipetting Guide
  • Reagent reservoir demo pack

Kit 3 includes:

  • Three Thermo Scientific Finnpipette Pipetters from 10 to 10000μL:
  • Finnpipette F1, 10-100μL
  • Finnpipette F1, 100-1000μL
  • Finnpipette F1, 1000-10000μL
  • Finntip Flex tips: 200, 1x96; 1000, 1x96; FT 10mL, 1x24 F-stand; F1 pen; F1 brochure
  • Good Laboratory Pipetting Guide
  • Reagent reservoir demo pack

Kit 4 includes:

  • Three Thermo Scientific Finnpipette Pipetters from 10 to 1000μL:
  • Finnpipette F1, 2-20μL
  • Finnpipette F1, 20-200μL
  • Finnpipette F1, 100-1000μL
  • Flex tips: (2) 200μL, 10x96; 100-1000μL, 10x96
  • F-stand; F1 pen; F1 brochure
  • Good Laboratory Pipetting Guide
  • Reagent reservoir demo pack

Ordering Information:

Good Laboratory Pipetting kit also includes samples of Finntip Flex for maximum performance and best ergonomics. The kit contains a Good Laboratory Pipetting guide offering a extensive information of pipetting and factors influencing to the pipetting results.

Compatible with:

Finntip, ART tip, HLT tip

TRUSTED_SUSTAINABILITY

Specifications

Compatible Tips Finntip Flex tips: 10; 200; 1000
Description Kit 1: Finnpipettes F1 1-10μL micro, 10-100μL and 100-1000μL
Color Assorted
For Use With Finntip: Flex Tips: 10, 200, 1000
Model F1
Product Line Finnpipette™ F1
Type GLP Kit
Volume (Metric) 1 to 1000 μL

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.