missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Finnpipette™ F1 Fixed Volume Single-Channel Pipettes

Product Code. 11827411
Click to view available options
Volume (Metric):
1 μL
10 μL
10,000 μL
100 μL
1000 μL
20 μL
200 μL
2000 μL
25 μL
250 μL
3000 μL
5 μL
50 μL
500 μL
5000 μL
Accuracy:
±0.40%
±0.70
+/-0.3%
+/-0.4%
+/-0.6%
+/-0.9%
+/-1.4%
+/-4%
Precision:
0.2%
0.3%
0.4%
0.5%
0.8%
1.4%
4%
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11827411

Brand: Thermo Scientific™ 4651080

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Protect samples from contamination with this manual pipette, featuring an adjustable finger rest and antimicrobial surface.

Protect samples from contamination with the Thermo Scientific™ Finnpipette™ F1 Fixed-Volume Pipette, featuring an adjustable finger rest, antimicrobial surface and super blow-out piston with volumes of 50μL and below. The Finnpipette F1 Fixed-Volume Pipette delivers light, smooth pipetting action with patented soft-touch tip ejection for maximum user comfort. The antimicrobial surface helps keep the pipette clean without the use of additional surface decontamination liquids. The entire pipette is resistant to UV light, reagents and moisture.

Features:
  • Antimicrobial surface: Silver (Ag) ions create a pipette surface that inhibits the growth of bacteria, microbes and fungus
  • Adjustable finger rest: Finger rest can be adjusted 120° for optimal pipetting position
  • Light, smooth pipetting action: Enables better results over longer pipetting periods
  • Super blow-out piston with volumes of 50μL and below: Ensures delivery of micro-size drops
  • Proprietary soft-touch tip ejection: Minimizes forces needed for tip ejection
TRUSTED_SUSTAINABILITY

Specifications

Accuracy +/-0.3%
Description F1 Fixed Volume Single Channel
Number of Channels 1
Type Fixed Volume Pipette
Volume (Metric) 1000 μL
Precision 0.3%

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.