missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Plastilab™ Finned Plug for Disposable Tubes

Product Code. 11361195
Change view
Click to view available options
Color:
Blue
Green
Neutral
Red
Yellow
Unit Size:
Pack of 1000
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
11361195 Green Pack of 1000
11341195 Yellow Pack of 1000
11655691 Red Pack of 1000
11615681 Neutral Pack of 1000
11331195 Blue Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11361195 Supplier Kartell™ Supplier No. 8830512

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Designed with flexible fins to create a snug fit.

  • Fits tubes 15 to 17mm 
  • Available in a variety of colors for coding

Specifications

Certifications/Compliance CE-IVD
Product Type Finned Plug
Material Polyethylene
Color Green
For Use With (Equipment) 15 to 17 mm Tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.