Learn More
BM Polyco™ Finite™ P Indigo AF Nitrile Powder Free Disposable Gloves
Product Code. p-8009690
Click to view available options
Size:
Small
Medium
Large
X-Large
Unit Size:
Pack of 100
Product Code. 12930685
Brand: BM Polyco™ MFNP100/02
Only null left
Add to basket
Edge
Add to basket
BM Polyco™ Finite™ P Indigo AF Nitrile Powder Free Disposable Gloves are fabricated from exceptionally soft and pliable material, and act as a barrier against oils, acids and chemicals, adhering to European Standard EN374
  • Advanced nitrile technology provides an exceptionally soft and pliable fit
  • Powder free and free from latex proteins and residual accelerators
  • Ideal for wearers with sensitive skin
  • Unique shape delivers an ambidextrous glove with improved fit
  • Barrier against oils, acids and chemicals adhering to European Standard EN374
  • Beaded cuffs offers additional strength whilst protecting the forearm from liquid roll back
  • Application areas: pharmaceuticals, electronics, inspection, laboratory work, clinical lab duties
Specifications
Certifications/Compliance European Standard EN374
Disposable Single-use
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size Medium
Sterility Non-sterile
Glove Cuff Style Beaded