missing translation for 'onlineSavingsMsg'
Learn More

Fingolimod Hydrochloride, Mikromol™

Product Code. 17944276
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17944276 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17944276 Supplier Mikromol Supplier No. MM3554.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Fingolimod Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fingolimod Hydrochloride
CAS 162359-56-0
Analyte or Component Names Fingolimod Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C19 H33 N O2 . Cl H
Certifications/Compliance ISO 17034
Synonym 2-Amino-2-[2-(4-octylphenyl)ethyl]-1,3-propanediol hydrochloride (1:1), 1,3-Propanediol, 2-amino-2-[2-(4-octylphenyl)ethyl]-, hydrochloride (9CI), 2-Amino-2-[2-(4-octylphenyl)ethyl]propane-1,3-diol hydrochloride, FTY 720, Fingolimod hydrochloride, Gilenya, Imusera
InChI Formula InChI=1S/C19H33NO2.ClH/c1-2-3-4-5-6-7-8-17-9-11-18(12-10-17)13-14-19(20,15-21)16-22;/h9-12,21-22H,2-8,13-16,20H2,1H3;1H
SMILES Cl.CCCCCCCCc1ccc(CCC(N)(CO)CO)cc1
IUPAC Name 2-amino-2-[2-(4-octylphenyl)ethyl]propane-1,3-diol;hydrochloride
Molecular Weight (g/mol) 343.93
Quantity 250 mg
Formula Weight 343.2278 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.