missing translation for 'onlineSavingsMsg'
Learn More

Ekastu Safety™ Mandil Filtering Facepiece FFP2/V with Cool Down Exhalation Valve

Product Code. 10155790
Change view
Click to view available options
Color:
Blue
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10155790 Blue Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10155790 Supplier Ekastu Safety™ Supplier No. 411281
Estimated Shipment: 23-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Protects against slightly toxic and harmful particles up to 12.5 times the NPF value. EKASTU Safety™ Mandil Filtering Facepiece FFP2/V with Cool Down Exhalation Valve has a five-year shelf life in original packaging.

  • With Cool Down exhalation valve
  • With colour-coded nosepiece and exhalation valve to immediate identification for the protection class
  • Protection against slightly toxic and harmful particles as well as unpleasant organic odours up to 12.5 times the NPF value
  • Activated charcoal layer included
  • Filter class DIN EN 149:2001 FFP2 NR
  • In original packaging: 5 years shelf life
  • 12 pieces per pack

Specifications

Color Blue
Filter Type FFP2
With Exhalation Valve Yes
Resistant To 160 l/min, constant flow max. 3.0 mbar
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.