missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ Finedex™ Latex Gloves

Product Code. 12605213
Click to view available options
Size:
9
10
Unit Size:
Each

Product Code. 12605213

Brand: Honeywell Safety Products™ 209440110

Only null left
Add to basket
Add to basket

Flexible latex, comfortable cotton flock, and a diamond-pattern finish make this glove a useful and practical choice for a variety of needs. Honeywell Safety Products™ Finedex™ Latex Gloves are manufactured via a process that minimizes latex protein.

  • Natural latex glove for flexibility, dexterity, and good mechanical and chemical resistance against detergents.
  • Cotton flock interior for comfort.
  • Diamond-pattern finish for grasping dry and damp objects.
  • Cuff protects forearm.
  • Minimum latex protein.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE CATEGORY II Intermediary risks, EN 420-2003 EEC Basic standards, EN 388-2003 Mechanical hazards, EN 374-2003 Chemical hazards, EC Category PPE 3, EC Certificate Number 0075/797/162/11/08/0462, EC Attestation Number 0075/797/162/11/08/0462
Classification Dexterity: 5
Color Yellow
Cut Resistance Rating 0
Exterior Finish Diamond
Material Latex
Product Type Gloves
Size 10
Thickness (Metric) 0.4 to 0.8 mm
For Use With (Application) Building, Construction, Catering, Chemical, Fishing, Food, Industrial Cleaning, Laboratory, Medical, Pharmaceutical, Printing, Ship Building and Water Treatment industries
Liner Material Cotton Flock
Lining Lined
Quantity 10Pair/PK
Tear Rating 1
Length (Metric) 30 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.