missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ FINEDEX™ 953-20 Nitrasoft Gloves

Product Code. 15896675
Change view
Click to view available options
Size:
7
8
9
Unit Size:
1 pair
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15896675 7 1 pair
15806685 8 1 pair
15816685 9 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15896675 Supplier Honeywell™ Supplier No. 209532007
Only null left
Add to basket
Add to basket

Honeywell™ FINEDEX™ 953-20 Nitrasoft Gloves designed for use in the agrifood industry (production of oils and fats, boning, packing of meat and fatty fish).

  • Unlined nitrile glove
  • Specific food formula for food industry
  • Chlorinated
  • Diamond pattern
  • Nitrile formula has been selected to give the glove greater elasticity and unrivalled dexterity
  • Special formula makes it suitable for the agrifood industry
  • Chlorinated interior makes the glove easier to slip on
  • Diamond finish makes the glove suitable for the grasping of dry and damp objects
  • The glove is particularly resistant to greases, oils and detergents
  • The cuff helps to protect a large part of the forearm
  • Treated to prevent the development of bacteria
  • It contains no natural latex or any allergenic protein
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EC Category PPE 3, EC Certificate 0075/797/162/11/08/0457, EC Attestation 0075/797/162/11/08/0457
Color Blue
Material Nitrile
Size 7
Thickness (Metric) 0.22 mm
Length (Metric) 33 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.