Learn More
Honeywell™ FINEDEX™ 944-01 Clean Gloves
Product Code. 12635213
Change view
Click to view available options
:
Size: 8
Product Code.
12635213 Size: 8
1 options
Product Code. 12635213 Supplier Honeywell™ Supplier No. 209440108
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Honeywell™ FINEDEX™ 944-01 Clean Gloves for general maintenance use in factories and collective areas.
  • Unlined yellow latex glove
  • Cotton flock inside
  • Diamond-pattern finish
  • Natural latex provides excellent flexibility, synonymous with dexterity and comfort
  • The cotton flocked interior provides the user with greater comfort
  • Diamond finish is tailored for grasping dry and damp objects
  • Latex offers good mechanical and chemical resistance against detergents and liquid products
  • The cuff helps to protect a large part of the forearm
  • The production process helps to obtain a glove with a minimum latex protein
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 420-2003 EEC Basic standards, EN 388-2003 Mechanical hazards, EN 374-2003 Chemical hazards
Color Yellow
Material Latex, Cotton
Size 8
Thickness (Metric) 0.4 to 0.8 mm
Length (Metric) 30 cm