Learn More
Epredia™ Fine Pore Cassettes
For very small biopsy tissue samples, cassettes have a front-edge lid hinge with attached lids that are available in 12 different color varieties
Brand: Epredia™ B851735LA
Additional Details : Weight : 4.53600kg
Description
Printing
- Surface finish and reinforced print faces are engineered to optimize print quality
- Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
- Cassette design and flow characteristics tested to ensure efficient reagent exchange
- Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
- Precision engineering for solid connection to microtomes
Archiving
- Cassette surface texture optimizes scannability for archiving
Specifications
Fine Pore | |
1000/Cs. | |
Attached Lid, Front Hinge | |
Lavender |
Yes | |
Loose Packed | |
No | |
Cassette |