missing translation for 'onlineSavingsMsg'
Learn More

Finasteride, MedChemExpress

Product Code. p-200098286
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
Unit Size:
100mg
200mg
This item is not returnable. View return policy

Product Code. 16486256

Brand: Medchem Express HY13635

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Finasteride (MK-906) is a potent and competitive 5α-reductase inhibitor, with an IC50 of 4.2 nM for type II 5α-reductase. Finasteride has approximately a 100-fold greater affinity for type II 5α-reductase enzyme than for the type I enzyme. Finasteride can be used for the research of benign prostatic hyperplasia (BPH) and androgenic alopecia.

Finasteride (MK-906) is a potent and competitive 5α-reductase inhibitor, with an IC50 of 4.2 nM for type II 5α-reductase. Finasteride has approximately a 100-fold greater affinity for type II 5α-reductase enzyme than for the type I enzyme. Finasteride can be used for the research of benign prostatic hyperplasia (BPH) and androgenic alopecia.

Specifications

Chemical Name or Material Finasteride
Physical Form Solid
CAS 98319-26-7
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C23H36N2O2
For Use With (Application) Cancer-programmed cell death
Synonym MK-906
Solubility Information DMSO : 150 mg/mL (402.64 mM; Need ultrasonic)
SMILES O=C([C@H]1CC[C@]2([H])[C@]1(C)CC[C@]3([H])[C@@]4(C)C=CC(N[C@]4([H])CC[C@]32[H])=O)NC(C)(C)C
Molecular Weight (g/mol) 372.54
Formula Weight 372.54
Percent Purity 98.73%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.