missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Finasteride, MedChemExpress
Click to view available options
Quantity:
100 mg
200 mg
Unit Size:
100mg
200mg
Description
Finasteride (MK-906) is a potent and competitive 5α-reductase inhibitor, with an IC50 of 4.2 nM for type II 5α-reductase. Finasteride has approximately a 100-fold greater affinity for type II 5α-reductase enzyme than for the type I enzyme. Finasteride can be used for the research of benign prostatic hyperplasia (BPH) and androgenic alopecia.
Specifications
Specifications
| Chemical Name or Material | Finasteride |
| Physical Form | Solid |
| CAS | 98319-26-7 |
| Color | White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C23H36N2O2 |
| For Use With (Application) | Cancer-programmed cell death |
| Synonym | MK-906 |
| Solubility Information | DMSO : 150 mg/mL (402.64 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction