missing translation for 'onlineSavingsMsg'
Learn More

Finasteride Acid, TRC

Product Code. 30135468
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135468 10 g 10g
17912255 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135468 Supplier Toronto Research Chemicals Supplier No. TRCO84725010G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104239-97-6
Molecular Formula C19 H27 N O3
Molecular Weight (g/mol) 317.42
Synonym 1H-Indeno[5,4-f]quinoline-7-carboxylic acid, 2,4a,4b,5,6,6a,7,8,9,9a,9b,10,11,11a-tetradecahydro-4a,6a-dimethyl-2-oxo-, (4aR,4bS,6aS,7S,9aS,9bS,11aR)-, 4-Azaandrost-1-ene-17-carboxylic acid, 3-oxo-, (5α,17β)-, (5α,17β)-N-[(2,5-Bis(trifluoromethyl)-phenyl]-3-oxo-4-aza-5-androst-1-ene-17-carboxylic acid, 1H-Indeno[5,4-f]quinoline-7-carboxylic acid, 2,4a,4b,5,6,6a,7,8,9,9a,9b,10,11,11a-tetradecahydro-4a,6a-dimethyl-2-oxo-, [4aR-(4aα,4bβ,6aα,7α,9aβ,9bα,11aβ)]-, 3-Oxo-4-aza-5α-androst-1-ene-17-carboxylic acid, 3-Oxo-4-aza-5α-androst-1-ene-17β-carboxylic acid, Dutasteride Imp A (EP)
IUPAC Name (1S,3aS,3bS,5aR,9aR,9bS,11aS)-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,9b,10,11-dodecahydroindeno[5,4-f]quinoline-1-carboxylic acid
SMILES C[C@]12CC[C@H]3[C@@H](CC[C@H]4NC(=O)C=C[C@]34C)[C@@H]1CC[C@@H]2C(=O)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C19H27NO3/c1-18-9-7-13-11(12(18)4-5-14(18)17(22)23)3-6-15-19(13,2)10-8-16(21)20-15/h8,10-15H,3-7,9H2,1-2H3,(H,20,21)(H,22,23)/t11-,12-,13-,14+,15+,18-,19+/m0/s1
Formula Weight 317.1991
Chemical Name or Material Finasteride acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.