missing translation for 'onlineSavingsMsg'
Learn More

Finasteride Acid, Mikromol™

Product Code. 17971746
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17971746 25 mg 25mg
17981046 100 mg 100mg
30463899 4 x 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17971746 Supplier Mikromol Supplier No. MM0415.050025
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Finasteride Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Finasteride acid
CAS 104239-97-6
Analyte or Component Names Finasteride acid
Shipping Condition Room Temperature
Molecular Formula C19 H27 N O3
Certifications/Compliance ISO 17025
Synonym 3-Oxo-4-aza-5alpha-alphandrost-1-ene-17beta-carboxylic Acid, 1H-Indeno[5,4-f]quinoline-7-carboxylic acid, 2,4a,4b,5,6,6a,7,8,9,9a,9b,10,11,11a-tetradecahydro-4a,6a-dimethyl-2-oxo-, (4aR,4bS,6aS,7S,9aS,9bS,11aR)-, 4-Azaandrost-1-ene-17-carboxylic acid, 3-oxo-, (5alpha,17beta)-, (5alpha,17beta)-N-[(2,5-Bis(trifluoromethyl)-phenyl]-3-oxo-4-aza-5-androst-1-ene-17-carboxylic acid, 1H-Indeno[5,4-f]quinoline-7-carboxylic acid, 2,4a,4b,5,6,6a,7,8,9,9a,9b,10,11,11a-tetradecahydro-4a,6a-dimethyl-2-oxo-, [4aR-(4aalpha,4bbeta,6aalpha,7alpha,9abeta,9balpha,11abeta)]-, 3-Oxo-4-aza-5alpha-androst-1-ene-17-carboxylic acid, 3-Oxo-4-aza-5alpha-androst-1-ene-17beta-carboxylic acid, Dutasteride Imp A (EP), Finasteride Acid, Dutasteride Acid Impurity, Dutasteride related, Dutasteride Impurity A
InChI Formula InChI=1S/C19H27NO3/c1-18-9-7-13-11(12(18)4-5-14(18)17(22)23)3-6-15-19(13,2)10-8-16(21)20-15/h8,10-15H,3-7,9H2,1-2H3,(H,20,21)(H,22,23)/t11-,12-,13-,14+,15+,18-,19+/m0/s1
SMILES C[C@]12CC[C@H]3[C@@H](CC[C@H]4NC(=O)C=C[C@]34C)[C@@H]1CC[C@@H]2C(=O)O
IUPAC Name (1S,3aS,3bS,5aR,9aR,9bS,11aS)-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,9b,10,11-dodecahydroindeno[5,4-f]quinoline-1-carboxylic acid
Molecular Weight (g/mol) 317.42
Quantity 25 mg
Formula Weight 317.1991 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.