missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenz Laborglasintrumente Final Receiver
Click to view available options
Capacity (Metric):
2 L
Unit Size:
Each
Description
- System contents and spare parts
- For one-way reaction/distillation units 6, 10, 20 l, LF 150
Specifications
Specifications
| Capacity (Metric) | 2 L |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction