missing translation for 'onlineSavingsMsg'
Learn More

FIN56, MedChemExpress

Product Code. 16464846
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16464846 100 mg 100mg
16446517 5 mg 5mg
16403798 10 mg 10mg
16402866 25 mg 25mg
16400177 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16464846 Supplier Medchem Express Supplier No. HY103087
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress FIN56 is a specific inducer of ferroptosis. FIN56 induces ferroptosis by inducing degradation of GPX4. FIN56 also binds to and activates squalene synthase.

FIN56 is a specific inducer of ferroptosis. FIN56 induces ferroptosis by inducing degradation of GPX4. FIN56 also binds to and activates squalene synthase.

Specifications

Chemical Name or Material FIN56
CAS 1083162-61-1
Color White
Purity Grade Notes Research
Molecular Formula C25H31N3O5S2
Quantity 100 mg
Solubility Information DMSO : ≥ 100 mg/mL (193.18 mM)
SMILES O=S(C1=CC(/C2=N\O)=C(C3=C2C=C(S(=O)(NC4CCCCC4)=O)C=C3)C=C1)(NC5CCCCC5)=O
Molecular Weight (g/mol) 517.66
Formula Weight 517.66
Percent Purity 98.17%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.