missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ FIN 56

Product Code. 18759286
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18759286 5 mg 5mg
18736296 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18759286 Supplier Tocris Bioscience™ Supplier No. 6280/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ferroptosis activator

FIN 56 is a bifunctional inducer of ferroptosis in vitro. Activates squalene synthase (SQS) leading to coenzyme Q10 depletion. Also induces degradation of glutathione peroxidase 4 (GPX4), via acetyl-CoA carboxylase (ACC).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N2,N7-Dicyclohexyl-9-(hydroxyimino)-9H-fluorene-2,7-disulfonamide
CAS 1083162-61-1
Assay Percent Range >98%
Quantity 5 mg
Target Ferroptosis Activators
Molecular Formula C25H31N3O5S2
Formula Weight Observed MW: 517.66
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.