missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ FIN 56
Description
FIN 56 is a bifunctional inducer of ferroptosis in vitro. Activates squalene synthase (SQS) leading to coenzyme Q10 depletion. Also induces degradation of glutathione peroxidase 4 (GPX4), via acetyl-CoA carboxylase (ACC).
Specifications
Specifications
| Chemical Name or Material | N2,N7-Dicyclohexyl-9-(hydroxyimino)-9H-fluorene-2,7-disulfonamide |
| CAS | 1083162-61-1 |
| Assay Percent Range | >98% |
| Quantity | 25 mg |
| Target | Ferroptosis Activators |
| Molecular Formula | C25H31N3O5S2 |
| Formula Weight | Observed MW: 517.66 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?