missing translation for 'onlineSavingsMsg'
Learn More

Fimepinostat, MedChemExpress

Product Code. 16455728
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16455728

Brand: Medchem Express HY13522

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Fimepinostat (CUDC-907) potently inhibits class I PI3Ks as well as classes I and II HDAC enzymes with an IC50 of 19/54/39 nM and 1.7/5.0/1.8/2.8 nM for PI3Kα/PI3Kβ/PI3Kδ and HDAC1/HDAC2/HDAC3/HDAC10 , respectively.

Fimepinostat (CUDC-907) potently inhibits class I PI3Ks as well as classes I and II HDAC enzymes with an IC50 of 19/54/39 nM and 1.7/5.0/1.8/2.8 nM for PI3Kα/PI3Kβ/PI3Kδ and HDAC1/HDAC2/HDAC3/HDAC10 , respectively.

Specifications

Chemical Name or Material Fimepinostat
CAS 1339928-25-4
Color White
Purity Grade Notes Research
Molecular Formula C23H24N8O4S
Quantity 10 mg
Synonym CUDC-907
Solubility Information DMSO : 50 mg/mL (98.32 mM; Need ultrasonic) ∣DMF : 5 mg/mL (9.83 mM; Need ultrasonic)
SMILES O=C(C1=CN=C(N(CC2=CC3=NC(C4=CC=C(OC)N=C4)=NC(N5CCOCC5)=C3S2)C)N=C1)NO
Molecular Weight (g/mol) 508.55
Formula Weight 508.55
Percent Purity 98.07%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.