missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toceranib, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Toceranib phosphate (SU11654 phosphate) is an orally active receptor tyrosine kinase (RTK) inhibitor, and it potently inhibits PDGFR, VEGFR, and Kit with Kis of 5 and 6 nM for PDGFRβ and Flk-1/KDR, respectively. Toceranib phosphate (SU11654 phosphate) has antitumor and antiangiogenic activity, and used in the treatment of canine mast cell tumors.
Specifications
Specifications
| Chemical Name or Material | Toceranib |
| CAS | 356068-94-5 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C22H25FN4O2 |
| Quantity | 50 mg |
| Synonym | SU11654 PHA 291639E |
| Solubility Information | DMSO : 2.5 mg/mL (6.31 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble) |
| SMILES | O=C(NCCN1CCCC1)C2=C(NC(/C=C3C(NC4=C\3C=C(C=C4)F)=O)=C2C)C |
| Molecular Weight (g/mol) | 396.46 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction