missing translation for 'onlineSavingsMsg'
Learn More

Fimasartan, TRC

Product Code. 30391027
Change view
Click to view available options
Quantity:
10 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30391027 250 mg
30391362 100 mg
30403537 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30391027 Supplier Toronto Research Chemicals Supplier No. TRCF342040250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 247257-48-3
Molecular Formula C27 H31 N7 O S
Molecular Weight (g/mol) 501.65
Synonym 5-Pyrimidineethanethioamide, 2-butyl-1,6-dihydro-N,N,4-trimethyl-6-oxo-1-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-, 5-Pyrimidineethanethioamide, 2-butyl-1,6-dihydro-N,N,4-trimethyl-6-oxo-1-[[2'-(1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]- (9CI), 2-Butyl-1,6-dihydro-N,N,4-trimethyl-6-oxo-1-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-5-pyrimidineethanethioamide, 2-n-Butyl-5-dimethylaminothiocarbonylmethyl-6-methyl-3-[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]pyrimidin-4(3H)-one, Fimasartan, Kanarb
IUPAC Name 2-[2-butyl-4-methyl-6-oxo-1-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]pyrimidin-5-yl]-N,N-dimethylethanethioamide
SMILES CCCCC1=NC(=C(CC(=S)N(C)C)C(=O)N1Cc2ccc(cc2)c3ccccc3c4nnn[nH]4)C

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C27H31N7OS/c1-5-6-11-24-28-18(2)23(16-25(36)33(3)4)27(35)34(24)17-19-12-14-20(15-13-19)21-9-7-8-10-22(21)26-29-31-32-30-26/h7-10,12-15H,5-6,11,16-17H2,1-4H3,(H,29,30,31,32)
Formula Weight 501.2311
Chemical Name or Material Fimasartan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.