missing translation for 'onlineSavingsMsg'
Learn More

GE Healthcare Nuclepore Polycarbonate Hydrophilic Membranes

Captures extremely small particles.

Brand:  GE Healthcare 113506

Code : 69

Additional Details : Weight : 0.18000kg

 View more versions of this product

Product Code. 11339004

  • £381.60 / Pack of 25


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Track-etched for highly defined pore size, high pore density
  • Treated with PVP to render it hydrophilic
  • Low extractables, minimizing sample contamination risk
  • High chemical resistance
  • Good thermal stability for wide range of sample types
  • Low, consistent ash and low tare weights
  • Smooth flat surface for high particle visibility
Specifications

Specifications

20.3 x 25.4cm
25/Pk.
0.2μm
Filtration Membranes
Hydrophilic
Sheet
Epifluorescence Microscopy, Cell Biology, EPA Testing
Polycarbonate
25Pack
Polycarbonate
Videos