Learn More
Witeg™ Filtration Flask
Product Code. 10477501
Change view
Click to view available options
Capacity: 2000 mL
Product Code.
10477501 Capacity: 2000 mL
1 options
Product Code. 10477501 Supplier Witeg™ Supplier No. 5 071 200
Only null left
Add to basket
Add to basket
In an Erlenmeyer shape to provide excellent mechanical strength under vacuum applications.
  • With PPN plastic hose connection and plastic tubature
  • With safety coating for extra protection and durability
  • Made of borosilicate glass
Specifications
Capacity (Metric) 2000 mL
Material Borosilicate Glass
For Use With (Application) Filtration, vacuum applications
Product Type Filtration flask
Shape Erlenmeyer
Bottom Shape Flat