Learn More
Integra Biosciences™ Pipetboy Filter
Product Code. 10531941
Change view
Click to view available options
Pore Size:
0.2 μm
0.45 μm
Sterility:
Non-sterile
Sterile
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Sterility unitSize
10531941 0.45 μm Sterile Each
10593425 0.2 μm Sterile Each
11656580 0.2 μm Non-sterile Each
10210143 0.45 μm Non-sterile Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10531941 Supplier Integra Biosciences™ Supplier No. 153015
Only null left
Add to basket
Edge
Add to basket
Prevent contamination with replaceable sterile filters. Integra Biosciences™ Pipetboy Filters are designed for use with Pipetboy Pro pipette controllers.
  • Spare filter
  • Accessory for Pipetboy Pro Pipette controllers
Specifications
Autoclavable Not Autoclavable
Pore Size 0.45 μm
Sterility Sterile
Type Filter
For Use With (Equipment) PipetBoy Pro pipette controller