missing translation for 'onlineSavingsMsg'
Learn More

Erlab Filtering Storage Cabinet Captair Smart

Product Code. 30245775
Change view
Click to view available options
Model:
1634
632B
834
Construction:
Pull-out Doors
Sliding Doors
Swing Doors
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Construction unitSize
30245775 834 Swing Doors Each
30245749 834 Pull-out Doors Each
30245766 1634 Swing Doors Each
30245774 1634 Pull-out Doors Each
30245758 632B Sliding Doors Each
30245772 632B Pull-out Doors Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 30245775 Supplier Erlab Supplier No. AB46//03

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

This filtering chemical storage cabinets Captair Smart provides outstanding filtration quality. We ensure users and laboratory technicians are protected against chemical hazards.

  • Safety Standards: Filtration performances tested according to the AFNOR NF X 15 211:2009 standard : France - EN 1822 : 1998 (HEPA H14 and ULPA U17 Filters)
  • CE Marking
  • Dimensions (mm): L 900 x D 660 x H 2140
  • Air Flow 220 m3/h / 129 CFM
  • Voltage / Frequency 110-230 V / 50-60 Hz
  • Power consumption 45 W
  • Structure: Corrosion resistant electro-galvanized steel coated with anti-acid polymer
  • Doors: Clear, chemical resistant acrylic for easy viewing
  • Filtration module is made with injected polypropylene
  • Filtration technology 1 column that can be configured to handle liquids, powders, or both
  • Following filtration column configuration: Carbon filtration for gases and vapors and particulate filtration for powders
  • Detection sensor for : Type S, for solvents / Type A, for acids / Type F, for formaldehydes
  • Temperature/Relative Humidity sensor
  • Capteur
  • Door sensors: Alarm if doors are left open
  • Storage compartments: 2
  • Delivered with 10 adjustable shelves with integrated retention tray
  • Absorbing mats: 2
  • Storage capacities: 100 x 1L glass bottles
  • Key lock
  • Communication Interface: When the light is pulsing: Doors left open - Containment is compromised - Filter breakthrough
  • eGuard APP: For remote control to monitor the status of the storage cabinet, change the settings, and deliver safety alerts immediately to your devices
  • Network connection

Specifications

Product Type Ductless Filtering Fume Hood
Filter Type Bonded Carbon and/or HEPA
Material Corrosion Resistant Electro-galvanized Steel, Antiacid Polymer Coating
Construction Swing Doors
Model 834
Voltage 110/230 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.