Learn More
Sartorius™ Filter Weighing Pan
Product Code. 15843075 Shop All Sartorius Products
Change view
Click to view available options
:
130 mm
Product Code.
15843075 130 mm
1 options
Product Code. 15843075 Supplier Sartorius™ Supplier No. YFW01SQP
Only null left
Add to basket
Add to basket
For use with Secura™ and Quintix™ lab balances.
  • 130 mm diameter
  • Accessory item
  • Conversion kit for 0.01 mg balances included
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Secura and Quintix Balance
For Use With (Application) Weighing
Size (Metric) 130 mm diameter
Type Weighing Pan