missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Filter Holder
Polysulfone In-Line Filter Holder for 50mm Diameter Membrane
Brand: Cytiva 10464100
This item is not returnable.
View return policy
Description
Polysulfone In-Line Filter Holder for 50mm Diameter Membrane

Specifications
Stainless Steel | |
ML050/0 Stainless Steel | |
1 Ea. | |
50 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction