Learn More
Agilent BioTek Filter Cubes for Imaging Multi-Mode Reader
For use with Imaging Multi-Mode Readers.
Brand: Agilent BioTek 1225110
Additional Details : Weight : 0.50000kg
Description
For use with Imaging Multi-Mode Readers.Order Info
Requires LED.
Specifications
Filter Cube | |
Imaging Multi-Mode Reader | |
CFP-YFP FRET |