missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Stedim Biotech™ Filter Boards, Grade C 450

Filter boards for the filtration and absorption of liquids.

Specifications

Basis Weight 450 g/m2
Format Sheet
Material Cotton Linters, Refined Pulp
Grade C 450
Quantity 100/Pk.
View More Specs

Products 2
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
15932770
View Documents Promotion Details
Sartorius Stedim Biotech™
FT-2-347-150200
150 mm × 200 mm N/A
View alternative products
Save Up to 
»
N/A
15942770
View Documents Promotion Details
Sartorius Stedim Biotech™
FT-2-347-580580
580 mm × 580 mm N/A
View alternative products
Save Up to 
»
N/A
Description

Description

  • Constructed of cotton linters and refined pulp
  • Suitable for analytical sample preparation
  • Non-sterile, bulk packed
Specifications

Specifications

450 g/m2
Cotton Linters, Refined Pulp
100/Pk.
Sheet
C 450
0.95 mm
Videos
SDS
Documents

Documents

Product Certifications