missing translation for 'onlineSavingsMsg'
Learn More

Filter aid, Harborlite 800

Product Code. 11313177
Change view
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2.5kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11313177 2500 g 2.5kg
11303177 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11313177 Supplier Thermo Scientific Alfa Aesar Supplier No. L13608.0E
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 130885-09-5

Filter aid, Harborlite 800 is used in multiple industrial applications such as chemical processing, mineral and lubrication oil production, biodiesel purification. It is also used for filtration performance in brewing, juices, wine, sugar syrups, edible vegetable and fish oils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Filter aid, Harborlite 800 is used in multiple industrial applications such as chemical processing, mineral and lubrication oil production, biodiesel purification. It is also used for filtration performance in brewing, juices, wine, sugar syrups, edible vegetable and fish oils.

Solubility
Insoluble in water.

Notes
Harborlite is a registered trademark of Harborlite Corportion.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130885-09-5
MDL Number MFCD00197888
Synonym Perlite, filter aid; Harborlite J2

Specifications

Density 2.3
Quantity 2500 g
Solubility Information Insoluble in water.
Chemical Name or Material Filter aid, Harborlite 800

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.