missing translation for 'onlineSavingsMsg'
Learn More

Filter aid, Celite Standard Super-cel

Product Code. 11449457
Change view
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449457 500 g 500g
11459457 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11449457 Supplier Thermo Scientific Alfa Aesar Supplier No. B21983.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 91053-39-3 | O2Si | 60.083 g/mol

Filter aid, Celite Standard Super-cel is used to maximize biodiesel yields as a result of improved filtration, dewatering, dewaxing and winterization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Filter aid, Celite Standard Super-cel is used to maximize biodiesel yields as a result of improved filtration, dewatering, dewaxing and winterization.

Solubility
Insoluble in water.

Notes
Incompatible with acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula O2Si
Molecular Weight (g/mol) 60.083
MDL Number MFCD00132803
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Odor Odorless
Quantity 500 g
CAS 61790-53-2
Merck Index 14,4973
Solubility Information Insoluble in water.
Chemical Name or Material Filter aid, Celite Standard Super-cel

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.