missing translation for 'onlineSavingsMsg'
Learn More

Filter Agent, Celite™ 545, Honeywell™ Fluka™

Catalog Number 15655480
Click to view available options
Quantity:
500 g
12 kg
Packaging:
Fiber Drum
Poly Bottle
This item is not returnable. View return policy
68855-54-9
CNa2O3
105.988
MFCD00132803
CDBYLPFSWZWCQE-UHFFFAOYSA-L
10340
CHEBI:29377
disodium;carbonate
C(=O)([O-])[O-].[Na+].[Na+]
This item is not returnable. View return policy
68855-54-9
CNa2O3
105.988
MFCD00132803
CDBYLPFSWZWCQE-UHFFFAOYSA-L
10340
CHEBI:29377
disodium;carbonate
C(=O)([O-])[O-].[Na+].[Na+]

Celite® 545

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Color White to Yellow
Quantity 500 g
Packaging Poly Bottle
UN Number NONH for all modes of transport
Chemical Name or Material Filter Agent
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'