missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Filter Agent, Celite™ 545, Honeywell™ Fluka™
Click to view available options
Quantity:
500 g
12 kg
Packaging:
Fiber Drum
Poly Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Color | White to Yellow |
Quantity | 12 kg |
Packaging | Fiber Drum |
UN Number | NONH for all modes of transport |
Chemical Name or Material | Filter Agent |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'