Learn More
3M™ DT-Series Filter
Product Code. 17883246
Change view
Click to view available options
Cartridge/Filter:
COMBINATION
PARTICULATE
Model:
CF32 A2B2E2K2P3 R D, DT-4045E
PF10 P3 R D, DT-1135E
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cartridge/Filter Model unitSize
17883246 COMBINATION CF32 A2B2E2K2P3 R D, DT-4045E Pack of 10
17863246 PARTICULATE PF10 P3 R D, DT-1135E Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17883246 Supplier 3M™ Supplier No. 7100260628
Add to basket
Add to basket
DT-Series filters provide effective protection against a range of particulates, gases and vapours, or combination hazards.
  • Includes specifications to provide protection against particulate hazards, gas & vapour hazards or a combination of both
  • Features a DIN40 mm threaded connection
  • Please note DT-Series filters were previously known as Pro2000 Filters
  • To identify which filter is suitable for your environment, and to aid in the implementation of a filter changeout schedule, please use the 3M Service Life and Select Software, available from www.3m.com/sls.
  • Can be used with certain 3M™ approved half and full-face tight-fitting respirators in negative-pressure mode
TRUSTED_SUSTAINABILITY
Specifications
Cartridge/Filter COMBINATION
Product Type Filter
Size Standard
For Use With (Equipment) 3M Air Purifying Respirators
Model CF32 A2B2E2K2P3 R D, DT-4045E