missing translation for 'onlineSavingsMsg'
Learn More

3M™ DT-Series Filter

Product Code. 17863246
Change view
Click to view available options
Cartridge/Filter:
COMBINATION
PARTICULATE
Model:
CF32 A2B2E2K2P3 R D, DT-4045E
PF10 P3 R D, DT-1135E
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cartridge/Filter Model unitSize
17863246 PARTICULATE PF10 P3 R D, DT-1135E Pack of 10
17883246 COMBINATION CF32 A2B2E2K2P3 R D, DT-4045E Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17863246 Supplier 3M™ Supplier No. 7100260622
Add to basket
Add to basket
This item is not returnable. View return policy

DT-Series filters provide effective protection against a range of particulates, gases and vapours, or combination hazards.

  • Includes specifications to provide protection against particulate hazards, gas & vapour hazards or a combination of both
  • Features a DIN40 mm threaded connection
  • Please note DT-Series filters were previously known as Pro2000 Filters
  • To identify which filter is suitable for your environment, and to aid in the implementation of a filter changeout schedule, please use the 3M Service Life and Select Software, available from www.3m.com/sls.
  • Can be used with certain 3M™ approved half and full-face tight-fitting respirators in negative-pressure mode
TRUSTED_SUSTAINABILITY

Specifications

Cartridge/Filter PARTICULATE
Color Black
Product Type Filter
Size Standard
For Use With (Equipment) 3M Air Purifying Respirators
Model PF10 P3 R D, DT-1135E
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.