missing translation for 'onlineSavingsMsg'
Learn More

Filorexant, MedChemExpress

Product Code. p-200098022
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16422807

Brand: Medchem Express HY15653

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Filorexant (MK-6096) is an orally bioavailable potent and selective reversible antagonist of OX1 and OX2 receptor(<3 nM in binding).

Filorexant (MK-6096) is an orally bioavailable potent and selective reversible antagonist of OX1 and OX2 receptor(<3 nM in binding).

Specifications

Chemical Name or Material Filorexant
CAS 1088991-73-4
Color Light Brown
Purity Grade Notes Research
Molecular Formula C24H25FN4O2
Quantity 50 mg
Synonym MK-6096
Solubility Information DMSO : 100 mg/mL (237.82 mM; Need ultrasonic)
SMILES CC1=CC(C(N2C[C@H](COC3=CC=C(F)C=N3)CC[C@H]2C)=O)=C(C4=NC=CC=N4)C=C1
Molecular Weight (g/mol) 420.48
Formula Weight 420.48
Percent Purity 98.95%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado