missing translation for 'onlineSavingsMsg'
Learn More

Filorexant, MedChemExpress

Product Code. 16422807
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16422807 50 mg 50mg
16419277 5 mg 5mg
16416866 100 mg 100mg
16406728 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16422807 Supplier Medchem Express Supplier No. HY15653
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Filorexant (MK-6096) is an orally bioavailable potent and selective reversible antagonist of OX1 and OX2 receptor(<3 nM in binding).

Filorexant (MK-6096) is an orally bioavailable potent and selective reversible antagonist of OX1 and OX2 receptor(<3 nM in binding).

Specifications

Chemical Name or Material Filorexant
CAS 1088991-73-4
Color Light Brown
Purity Grade Notes Research
Molecular Formula C24H25FN4O2
Quantity 50 mg
Synonym MK-6096
Solubility Information DMSO : 100 mg/mL (237.82 mM; Need ultrasonic)
SMILES CC1=CC(C(N2C[C@H](COC3=CC=C(F)C=N3)CC[C@H]2C)=O)=C(C4=NC=CC=N4)C=C1
Molecular Weight (g/mol) 420.48
Formula Weight 420.48
Percent Purity 98.95%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.