Learn More
Beckman Coulter Filling Solution, 4 M KCl Saturated with AgCl forAgCl electrodes (4-pack of 100 mL)
Brand:  Beckman Coulter 566467
Encompass
Filling Solution, 4 M KCl Saturated with AgCl forAgCl electrodes (4-pack of 100 mL)
Product Code. 10579250