missing translation for 'onlineSavingsMsg'
Learn More

Filipin III, TRC

Product Code. 30398294
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398294 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30398294 Supplier Toronto Research Chemicals Supplier No. TRCF3370031MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Filipin III
CAS 480-49-9
Molecular Formula C35H58O11
Quantity 1 mg
Synonym Oxacyclooctacosane, Filipin III deriv., 14-Deoxylagosin, 15-Deoxylagosin, 1',26-Dihydroxyfilipin I, 1'-Hydroxyfilipin II, 1'-epi-Filipin IV, Filimarisin, NSC 3364, NSC 38092, [3R-[3R*(R*),4S*,6S*,8S*,10R*,12R*,14R*,16S*,17E,19E,21E,23E,25E,27S*,28R*]]-4,6,8,10,12,14,16,27-Octahydroxy-3-(1-hydroxyhexyl)-17,28-dimethyloxacyclooctacosa-17,19,21,23,25-pentaen-2-one
InChI Formula InChI=1S/C35H58O11/c1-4-5-11-16-31(42)34-33(44)22-29(40)20-27(38)18-25(36)17-26(37)19-28(39)21-32(43)23(2)14-12-9-7-6-8-10-13-15-30(41)24(3)46-35(34)45/h6-10,12-15,24-34,36-44H,4-5,11,16-22H2,1-3H3/b7-6+,10-8+,12-9+,15-13+,23-14?/t24-,25+,26-,27+,28-,29+,30+,31-,32+,33+,34-/m1/s1
SMILES CCCCC[C@@H](O)[C@@H]1[C@@H](O)C[C@@H](O)C[C@@H](O)C[C@@H](O)C[C@@H](O)C[C@@H](O)C[C@H](O)C(=C\C=C\C=C\C=C\C=C\[C@H](O)[C@@H](C)OC1=O)C
IUPAC Name (3R,4S,6S,8S,10R,12R,14R,16S,19E,21E,23E,25E,27S,28R)-4,6,8,10,12,14,16,27-octahydroxy-3-[(1R)-1-hydroxyhexyl]-17,28-dimethyl-1-oxacyclooctacosa-17,19,21,23,25-pentaen-2-one
Molecular Weight (g/mol) 654.83
Formula Weight 654.3979
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.