missing translation for 'onlineSavingsMsg'
Learn More

Filgotinib, MedChemExpress

Product Code. p-200095805
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16483517 50 mg 50mg
16479977 5 mg 5mg
16457498 10 mg 10mg
16403606 25 mg 25mg
16477456 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16483517 Supplier Medchem Express Supplier No. HY18300

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Filgotinib (GLPG0634) is a selective and orally active JAK1 inhibitor with IC50 of 10 nM, 28 nM, 810 nM, and 116 nM for JAK1, JAK2, JAK3, and TYK2, respectively.

Filgotinib (GLPG0634) is a selective and orally active JAK1 inhibitor with IC50 of 10 nM, 28 nM, 810 nM, and 116 nM for JAK1, JAK2, JAK3, and TYK2, respectively.

Specifications

Chemical Name or Material Filgotinib
CAS 1206161-97-8
Color White
Purity Grade Notes Research
Molecular Formula C21H23N5O3S
Quantity 50 mg
Synonym GLPG0634
Solubility Information DMSO : 25 mg/mL (58.75 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1CC1)NC2=NN3C(C4=CC=C(CN5CCS(CC5)(=O)=O)C=C4)=CC=CC3=N2
Molecular Weight (g/mol) 425.5
Formula Weight 425.5
Percent Purity 98.08%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.