missing translation for 'onlineSavingsMsg'
Learn More

Filensin Antibody (6B4), Novus Biologicals™

Product Code. 18371538 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18371538 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18371538 Supplier Novus Biologicals Supplier No. H00000631M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Filensin Monoclonal antibody specifically detects Filensin in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Filensin
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 6B4
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_001186
Gene Alias Beaded filament structural protein 1, beaded filament structural protein 1, filensin, CP115Lens intermediate filament-like heavy, filensin, Lens fiber cell beaded-filament structural protein CP 115, LIFL-H115 KD
Host Species Mouse
Immunogen BFSP1 (NP_001186, 567 a.a. ~ 664 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. EESRRPCAMVTPGAEEPSIPEPPKPAADQDGAEVLGTRSRSLPEKGPPKALAYKTVEVVESIEKISTESIQTYEETAVIVETMIGKTKSDKKKSGEKS
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 631
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.