Learn More
Filamin B Antibody (1372), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-43732
Product Code. 18133379
  • £494.00 / 0.10mL
Description
Ensure accurate, reproducible results in Western Blot
Filamin B Monoclonal specifically detects Filamin B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Filamin B
Monoclonal
Unconjugated
PBS with No Preservative
FLNB
Recombinant protein encompassing a sequence within the N-terminus region of human Filamin B. The exact sequence is proprietary.
Protein G purified
RUO
2317
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
IgG2a
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
1372
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunoprecipitation 1:100-1:500, Immunohistochemistry-Paraffin 1:100-1:1000
ABP-278filamin B, beta (actin binding protein 278), ABP-280 homolog, actin binding protein 278, Actin-binding-like protein, AOI, Beta-filamin, DKFZp686O033, Fh1, filamin B, beta, Filamin homolog 1, filamin-3, filamin-B, FLN1L, FLN3, FLN-B, Larsen syndrome 1 (autosomal dominant), LRS1, SCT, TABPABP-280, TAPDKFZp686A1668, Thyroid autoantigen, Truncated ABP, Truncated actin-binding protein
Mouse
278 kDa
0.1 mL
Primary
Human, Mouse, Rat
Purified