missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Filamin A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£292.00 - £433.00
Specifications
| Antigen | Filamin A |
|---|---|
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Research Discipline | Neuroscience, Phospho Specific |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18652311
|
Novus Biologicals
NBP2-76488-25ul |
25 μL |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18376441
|
Novus Biologicals
NBP2-76488 |
100 μL |
£433.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Filamin A Polyclonal specifically detects Filamin A in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| Filamin A | |
| Unconjugated | |
| Neuroscience, Phospho Specific | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
| 2316.0 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: FQVTALAGDQPSVQPPLRSQQLAPQYTYAQGGQQTWAPERPLVGVNGLDVTSLRPFDLVIPFTIKKGEITGEVRMPSGKVAQPTITDNKDGTVTVRYAPSEAGLHEMDIRYDNMHIPGS | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Polyclonal | |
| Rabbit | |
| Human | |
| ABP-280DKFZp434P031, ABPX, actin binding protein 280, Actin-binding protein 280, Alpha-filamin, CVD1, EC 2.1.1.43, EC 6.3.4.4, Endothelial actin-binding protein, filamin A, alpha, filamin A, alpha (actin binding protein 280), Filamin-1, filamin-A, FLN, FLN1OPD, FLN-A, FMD, MNS, NHBP, Non-muscle filamin, OPD1, OPD2, XLVD, XMVD | |
| FLNA | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title