missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Filamin 2 Polyclonal Antibody

Filamin 2 Polyclonal Antibody
Brand: Invitrogen PA588919
This item is not returnable.
View return policy
Description
Filamin 2 Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| Filamin 2 | |
| Polyclonal | |
| Unconjugated | |
| FLNC | |
| 1110055E19Rik; ABP-280; ABP280A; ABP-280-like protein; ABPA; ABPL; ABP-L; ABP-L, gamma filamin; actin binding protein 280; actin-binding protein 280; actin-binding-like protein; filamin C; Filamin C gamma; filamin C, gamma; filamin C, gamma (actin binding protein 280); filamin, gamma; filamin-2; Filamin-C; FLJ10186; FLN2; FLNC; FLN-C; gamma filamin; gamma-filamin; MFM5; MPD4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 2318 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.01 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q14315 | |
| FLNC | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 2160-2340 of human FLNC (NP_0014493). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction