missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Filaggrin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583129
This item is not returnable.
View return policy
Description
Immunogen sequence: SADSSRQSGT PHAETSSGGQ AASSHEQARS SPGERHGSRH QQSADSSRHS GIPRRQASSA VRDSGHWGSS GSQASDSEGH SEESDTQSVS GHGQDGPHQQ SHQESAR Highest antigen sequence indentity to the following orthologs: Rat - 36%, Mouse - 38%.
Filaggrin is expressed only in well differentiated keratinized epithelial cells. The synthesis of filaggrin varies in types of ichthyosis, and with viral, premalignant and malignant conditions.
Specifications
| Filaggrin | |
| Polyclonal | |
| Unconjugated | |
| FLG | |
| ATOD2; AW107830; epidermal filaggrin; filaggrin; Filaggrin (profilaggrin); FLG; ft; Pflg | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2312 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P20930 | |
| FLG | |
| Recombinant protein corresponding to Human FLG. Recombinant protein control fragment (Product #RP-94296). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction